Merck
  • Diethylcarbamazine citrate ameliorates insulin resistance in high-fat diet-induced obese mice via modulation of adipose tissue inflammation.
Diethylcarbamazine citrate ameliorates insulin resistance in high-fat diet-induced obese mice via modulation of adipose tissue inflammation.
International immunopharmacology (2015-10-04)
Mahmoud Abdel-Latif
ABSTRAKT
Diethylcarbamazine citrate (DEC) had been known as anti-inflammatory drug but its effect on obesity-induced insulin resistance as a result of released inflammatory mediators from adipose tissue (AT) was not known. White male albino mice were fed with high fat diet (HFD) for 18weeks to induce obesity. DEC at different three doses (12, 50 and 200mg/kg) was orally administered twice a week starting at week 6. Body, liver and adipose tissue weights were taken, while glucose tolerance, insulin resistance, blood triglycerides and levels of adipokines (leptin, TNF-α, IL-6 and MCP-1) were tested. The activity of cyclooxygenase (COX) in the liver tissue homogenate was also tested. In addition, NF-κBp65 localization in liver cell cytoplasmic and nuclear fractions was detected using Western blotting. The only effective anti-inflammatory dose was 50mg/kg to reduce (p<0.05) the high levels of glucose, insulin and triglycerides in serum. DEC was not anti-obesity drug because the weights of body, liver and adipose tissues were not changed. Hyperleptinemia was decreased (p<0.001) and associated with a reduction in serum levels of TNF-α, IL-6 and MCP-1 (p<0.001). In addition, the activity of COX in DEC treatment decreased significantly (p<0.01), while NF-κBp65 localization in nuclear extracts was obviously inhibited in 50mg/kg treated group. It could be concluded that DEC was the only effective dose against mouse insulin resistance but not lipid accumulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hematin porcine