Merck
Hepatitis C virus non-structural protein 4 suppresses Th1 responses by stimulating IL-10 production from monocytes.
European journal of immunology (2003-11-25)
Miriam T Brady, Angus J MacDonald, Aileen G Rowan, Kingston H G Mills
ABSTRAKT
The majority of hepatitis C virus (HCV) infections become chronic, despite the presence of HCV-specific cellular and humoral immune responses. We have previously suggested that IL-10-secreting antigen-specific regulatory T cells may contribute to viral persistence, and demonstrate here that peripheral blood mononuclear cells (PBMC) from chronically HCV-infected patients secrete IL-10, but not IFN-gamma, in response to HCV nonstructural protein 4 (NS4). A neutralizing anti-IL-10 antibody restored this defective antigen-specific IFN-gamma production in vitro. Furthermore, PBMC from normal individuals secreted IL-10 in response to NS4, suggesting that cells of the innate immune system, in addition to T cells, produced IL-10 in the HCV-infected patients. Cell separation experiments revealed that the innate IL-10 was produced by blood monocytes, but not dendritic cells (DC). In addition, NS4 inhibited IL-12 production by PBMC in response to LPS and IFN-gamma, and Th1 responses to recall antigens in normal individuals. Furthermore, supernatants from NS4-stimulated monocytes inhibited LPS-induced maturation of DC and suppressed their capacity to stimulate proliferation and IFN-gamma production by allospecific T cells. Our data suggest that HCV subverts cellular immunity by inducing IL-10 and inhibiting IL-12 production by monocytes, which in turn inhibits the activation of DC that drive the differentiation of Th1 cells.