Merck
Comparisons of biophysical properties and bioactivities of mono-PEGylated endostatin and an endostatin analog.
Chinese journal of cancer (2016-01-23)
Shan Wang, Yan Fu, Yongzhang Luo
ABSTRAKT
Endostatin (ES) is a well-established potent endogenous antiangiogenic factor. An ES variant, called zinc-binding protein-ES (ZBP-ES), is clinically available; however, its use is limited by rapid renal clearance and short residence time. PEGylation has been exploited to overcome these shortcomings, and mono-PEGylated ES (called M2ES) as well as mono-PEGylated ZBP-ES (MZBP-ES) are developed in our study. This study aimed to compare the biophysical properties and biological effects of M2ES and MZBP-ES to evaluate their druggability. Circular dichroism and tryptophan emission fluorescence were used to monitor the conformational changes of M2ES and MZBP-ES. Their resistance to trypsin digestion and guanidinium chloride (GdmCl)-induced unfolding was examined by Coomassie staining and tryptophan emission fluorescence, respectively. The biological effects of M2ES and MZBP-ES on endothelial cell migration were evaluated using Transwell migration and wound healing assays, and the uptake of M2ES and MZBP-ES in endothelial cells was also compared by Western blotting and immunofluorescence. Structural analyses revealed that M2ES has a more compact tertiary structure than MZBP-ES. Moreover, M2ES was more resistant to trypsin digestion and GdmCl-induced unfolding compared with MZBP-ES. In addition, although M2ES and MZBP-ES showed comparable levels of inhibiting transwell migration and wound healing of endothelial cells, M2ES displayed an increased ability to enter cells compared with MZBP-ES, possibly caused by the enhanced interaction with nucleolin. M2ES has a more compact tertiary structure, is more stable for trypsin digestion and GdmCl-induced unfolding, exhibits increased cellular uptake and shows equivalent inhibitory effects on cell migration relative to MZBP-ES, indicating that M2ES is a more promising candidate for anticancer drug development compared with MZBP-ES.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O