Merck
A novel histone deacetylase 1 and 2 isoform-specific inhibitor alleviates experimental Parkinson's disease.
Neurobiology of aging (2015-11-08)
Chi-Jing Choong, Tsutomu Sasaki, Hideki Hayakawa, Toru Yasuda, Kousuke Baba, Yoshiyuki Hirata, Shinichi Uesato, Hideki Mochizuki
ABSTRAKT
With increased histone deacetylase (HDAC) activity and histone hypoacetylation being implicated in neurodegeneration, HDAC inhibitors have been reported to have considerable therapeutic potential. Yet, existing inhibitors lack specificity and may show substantial adverse effect. In this study, we identified a novel HDAC1/2 isoform-specific inhibitor, K560, with protective effects against 1-methyl-4-phenylpyridinium (MPP(+))- and/or 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-induced neuronal death in both in vitro and in vivo Parkinson's disease model. K560 attenuated cell death induced by MPP(+) in differentiated SH-SY5Y cells through the sustained expression of an antiapoptotic protein, X-linked inhibitor of apoptosis (XIAP). Inhibition of XIAP expression by locked nucleic acid antisense oligonucleotides abolished the protective effect of K560. Inactivation of mitogen-activated protein kinase cascades, reduced p53 phosphorylation, and down-regulation of p53-upregulated modulator of apoptosis on K560 treatment were also observed. Furthermore, pre- and post-oral administration of K560 to mice prevented MPTP-induced loss of dopaminergic neurons in substantia nigra, suggesting that selective inhibition of HDAC1 and HDAC2 by K560 may pave the way to new strategies for Parkinson's disease treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Apicidin, ≥98% (HPLC), from microbial
Sigma-Aldrich
SAHA, ≥98% (HPLC)