Merck
Extracorporeal shock wave therapy effectively prevented diabetic neuropathy.
American journal of translational research (2016-02-18)
Yi-Ling Chen, Kuan-Hung Chen, Tsung-Cheng Yin, Tien-Hung Huang, Chun-Man Yuen, Sheng-Ying Chung, Pei-Hsun Sung, Meng-Shen Tong, Chih-Hung Chen, Hsueh-Wen Chang, Kun-Chen Lin, Sheung-Fat Ko, Hon-Kan Yip
ABSTRAKT
We tested the hypothesis that extracorporeal shock wave (ECSW) therapy can effectively protect sciatic nerve (SN) from diabetes mellitus (DM)-induced neuropathy in leptin-deficient (ob/ob) mice. Eighteen-week C57BL/6 mice (n=8) served as age-matched controls (group 1) and ob/ob mice (n=16) were categorized into DM (group 2) and DM + ECSW (0.12 mJ/mm(2) for 4 times of 200 impulses at 3-week intervals) (group 3). The animals were sacrificed two weeks post-ECSW. In vitro results showed that the protein expressions of oxidative stress (NOX-1, NOX-2, oxidized protein), inflammation (MMP-9, TNF-α, iNOS), apoptosis (Bax, cleaved caspase-3, & PARP), and DNA-damage marker (γ-H2AX) were significantly higher in RT4-D6P2T (schwannoma cell line) treated by menadione (25 µM) compared with control group and were significantly reversed after ECSW (0.12 mJ/mm(2), 200 impulses) (all p<0.001). mRNA expressions of inflammation (MMP-9, TNF-α, iNOS), oxidative stress (NOX-1, NOX-2) and apoptosis (Bax, caspase-3) in SN were significantly higher in group 2 than in group 1 and were significantly reversed in group 3, whereas the mRNA expressions of anti-oxidants (HO-1, NQO1) progressively increased from group 1 to group 3 (all p<0.001). Cellular expressions of F4/80+, CD14+, γ-H2AX+ cells, and number of vacuolar formation in SN showed a pattern identical to that of inflammation markers among all groups (all p<0.001). Microscopic findings of Schwann cells and myelin-sheath scores, and number of eNOS+ cells in SN showed a reversed pattern compared to that of inflammation among all groups (all p<0.001). ECSW therapy protected SN against DM-induced neuropathy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%