Merck
Sex-Based Differences in Perceived Pragmatic Communication Ability of Adults With Traumatic Brain Injury.
Archives of physical medicine and rehabilitation (2015-02-11)
Emily H Despins, Lyn S Turkstra, Margaret A Struchen, Allison N Clark
ABSTRAKT
To identify sex-based differences in self-reported and close other-reported perceptions of communication behaviors in adults with traumatic brain injury (TBI). Between-groups comparison of questionnaire data from men and women with TBI and their close others. University academic department. Adults with medically documented TBI (n=160) and adults without TBI (n=81; control group) (N=241). Not applicable. La Trobe Communication Questionnaire, a standardized measure of communication problems in everyday life. Participants with TBI endorsed more communication problems than controls (P<.001). There were no significant differences in self-ratings (P=.20) or in the ratings of close others (P=.09) in communication behaviors of men with TBI compared with women with TBI. There was no difference between the self-ratings of women with TBI and their close others (P=.59). However, men with TBI significantly underreported communication problems compared with reports of close others (P<.001). Women with TBI might be more accurate than men with TBI in recognizing their own pragmatic communication problems.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%