Merck
  • Presence of cathepsin B in the human pancreatic secretory pathway and its role in trypsinogen activation during hereditary pancreatitis.
Presence of cathepsin B in the human pancreatic secretory pathway and its role in trypsinogen activation during hereditary pancreatitis.
The Journal of biological chemistry (2002-04-05)
Zoltán Kukor, Julia Mayerle, Burkhard Krüger, Miklós Tóth, Paul M Steed, Walter Halangk, Markus M Lerch, Miklós Sahin-Tóth
ABSTRAKT
The lysosomal cysteine protease cathepsin B is thought to play a central role in intrapancreatic trypsinogen activation and the onset of experimental pancreatitis. Recent in vitro studies have suggested that this mechanism might be of pathophysiological relevance in hereditary pancreatitis, a human inborn disorder associated with mutations in the cationic trypsinogen gene. In the present study evidence is presented that cathepsin B is abundantly present in the secretory compartment of the human exocrine pancreas, as judged by immunogold electron microscopy. Moreover, pro-cathepsin B and mature cathepsin B are both secreted together with trypsinogen and active trypsin into the pancreatic juice of patients with sporadic pancreatitis or hereditary pancreatitis. Finally, cathepsin B- catalyzed activation of recombinant human cationic trypsinogen with hereditary pancreatitis-associated mutations N29I, N29T, or R122H were characterized. In contrast to a previous report, cathepsin B-mediated activation of wild type and all three mutant trypsinogen forms was essentially identical under a wide range of experimental conditions. These observations confirm the presence of active cathepsin B in the human pancreatic secretory pathway and are consistent with the notion that cathepsin B-mediated trypsinogen activation might play a pathogenic role in human pancreatitis. On the other hand, the results clearly demonstrate that hereditary pancreatitis-associated mutations do not lead to increased or decreased trypsinogen activation by cathepsin B. Therefore, mutation-dependent alterations in cathepsin B-induced trypsinogen activation are not the cause of hereditary pancreatitis.