- Soluble polymer bound cleavage reagents: a multipolymer strategy for the cleavage of tertiary amines from REM resin.
Soluble polymer bound cleavage reagents: a multipolymer strategy for the cleavage of tertiary amines from REM resin.
Organic letters (2000-08-10)
P H Toy, T S Reger, K D Janda
PMID10930244
ABSTRAKT
Soluble polymer bound reagent 1 has been prepared to cleave tertiary amines from REM resin. Normally, amines cleaved from REM resin require extraction or chromatography to remove excess cleavage reagent and its byproducts. The solubility profile of non-crosslinked polystyrene (NCPS) based reagent 1 eliminates the need for such purification and allows for the direct isolation of a library of pure tertiary amines through simple filtration and concentration operations.