Merck
Fatsioside A‑induced apoptotic death of HepG2 cells requires activation of AMP‑activated protein kinase.
Molecular medicine reports (2015-08-08)
Yu-Shan Zheng, Jian-You Zhang, Dong-Hui Zhang
ABSTRAKT
Hepatocellular carcinoma (HCC) is one of the most malignant types of human primary tumor and has a poor prognosis, therefore, the development of novel therapeutic modalities is necessary. Fatsioside A is a novel baccharane‑type triterpenoid glycoside, which is extracted from the fruits of Fatsia japonica. Previous data has revealed that fatsioside A can exert growth inhibition, cell cycle arrest and induce apoptosis in human glioma cells. However, no detailed investigations have been performed to determine its action on human hepatocellular cells, and the exact mechanisms underlying the induction of apoptosis remain to be elucidated. The aim of the present study was to investigate the anticancer effect of fatsioside A in the HepG2 human HCC cell line, and to investigate the underlying mechanisms by focusing on the AMP‑activated protein kinase (AMPK) signaling cascade. The results of the present study demonstrated that fatsioside A induced apoptotic death of the human HepG2 HCC cells, which was associated with a marked activation of AMPK and increased expression of the downstream acetyl‑CoA carboxylase carboxylase. Inhibition of AMPK by RNA interference or by its inhibitor, compound C, suppressed fatsioside A‑induced caspase‑3 cleavage and apoptosis in the HepG2 cells, while AICAR, the AMPK activator, elicited marked cytotoxic effects. Together, these results suggested that fatsioside A‑induced apoptotic death requires AMPK activation in HepG2 cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Inhibitor AMPK, związek C, AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC50 = 25.1, 148, and 234.6 nM, respectively).
Sigma-Aldrich
Caspase Inhibitor VI, Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.
Sigma-Aldrich
Anti-Bcl-2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-AMPK α (pThr172) antibody produced in rabbit, affinity isolated antibody