Merck
Profiling of hepatic clearance pathways of Pittsburgh compound B and human liver cytochrome p450 phenotyping.
EJNMMI research (2013-02-15)
Anne Van Vlaslaer, Russell J Mortishire-Smith, Claire Mackie, Xavier Langlois, Mark E Schmidt
ABSTRAKT
11C-PiB has been developed as a positron-emission tomography (PET) ligand for evaluating fibrillar β-amyloid (Aβ) in the human brain. The ligand is rapidly metabolized, with approximately 10% of intact tracer remaining 30 min after injection. When 11C-PiB is used as a treatment endpoint in intervention studies for Alzheimer's disease (AD), a concern is whether the clearance of the tracer changes from one scan to the next, increasing within subject variability in the PET signal. Subjects enrolled in AD trials may start or stop medications that inhibit or induce xenobiotic metabolizing enzymes such as the cytochrome P450 (CYP) isozymes. We conducted CYP phenotyping in recombinantly expressed systems, and in human liver microsomes, to evaluate CYP isozyme contributions to the metabolism of PiB (carrier) and profiled microsomal and hepatocyte incubations for metabolites. The metabolism of PiB appears to be polyzymic, with direct conjugation via UDP-glucuronosyltransferases (UGTs) also occurring. It is unlikely that CYP inhibition or induction will significantly influence the clearance of 11C-PiB.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Naphthoflavone, ≥98%