Merck
Combination of SLC administration and Tregs depletion is an attractive strategy for targeting hepatocellular carcinoma.
Molecular cancer (2013-12-07)
Long Chen, Shuang Zhou, Jie Qin, Heng Hu, Huiying Ma, Binbin Liu, Xuan Wang, Jiaqi Ma, Shenglong Ye, Cuiping Zhong, Guomin Zhou, Chunmin Liang
ABSTRAKT
Secondary lymphoid tissue chemokine (SLC) is a key CC chemokine for chemotaxis of immune cells and has been an attractive candidate for anti-tumor treatments. However, among the immune cells recruited by SLC to tumors, the CD25+ Foxp3+ regulatory T cells (Tregs) compromise the anti-tumor effects. In this study, we proposed the combination therapy of intratumoral co-administration of SLC and anti-CD25 monoclonal antibodies (mAbs). We hypothesized that the intratumoral injections of SLC and depletion of Tregs would have stronger inhibition effects on the progression of hepatocellular carcinoma (HCC) in mice. C57BL/6 mice were inoculated subcutaneously with the murine HCC cell line, and mice with visible tumors were treated intratumorally with SLC, SLC plus anti-CD25 mAbs or the control antibodies. The percentages of Tregs, effector CD8+ T cells and CD4+ T cells were checked in the tumors, lymph nodes, spleen and liver at regular intervals. The levels of intratumoral IL-12, IFN-γ, IL-10 and TGF-β1 were evaluated. The final anti-tumor effects were measured by the tumor volume and weight as well as the intratumoral activity of MMP2 and MMP9. Bone-marrow-derived dendritic cells were used to explore the mechanisms of maturation induced by SLC in vitro. Our experiments showed the combination therapy significantly decreased the frequency of Tregs, and increased CD8+ T cells and CD4+ T cells at tumor sites. These alterations were accompanied by an increased level of IL-12 and IFN-γ, and decreased level of IL-10 and TGF-β1. Unexpectedly, we observed a significantly decreased percentage of Tregs, and increased CD8+ T cells and CD4+ T cells in the lymph nodes, spleen and liver after the combination therapy. The growth and invasiveness of HCC was also maximally inhibited in the combination therapy compared with the SLC alone. Furthermore, we confirmed SLC induced the maturation of DCs via NF-κB p65 and this maturation would benefit the combination therapy. Our data demonstrated that intratumoral co-administration of SLC and anti-CD25 mAbs was an effective treatment for HCC, which was correlated with the altered tumor microenvironment and systemically optimized percentages of Tregs, CD8+ T cells and CD4+ T cells in peripheral immune organs.