Merck
  • A general method for synthesis of trifluoroacetyltrialkyl (aryl) silanes and the Sakurai reaction of fluorinated acylsilanes with allyl silanes
A general method for synthesis of trifluoroacetyltrialkyl (aryl) silanes and the Sakurai reaction of fluorinated acylsilanes with allyl silanes
Journal of Fluorine Chemistry (2004)
Chung WJ and Welch JT