Merck
Skeletal Muscle Remodelling as a Function of Disease Progression in Amyotrophic Lateral Sclerosis.
BioMed research international (2016-05-20)
L Jensen, L H Jørgensen, R D Bech, U Frandsen, H D Schrøder
ABSTRAKT
Muscle weakness is considered the pivotal sign of amyotrophic lateral sclerosis (ALS). Knowledge about the skeletal muscle degeneration/regeneration process and the myogenic potential is limited in ALS patients. Therefore, we investigate these processes in a time course perspective by analysing skeletal muscle biopsies from ALS patients collected before and after a 12-week period of normal daily activities and compare these with healthy age-matched control tissue. We do this by evaluating mRNA and protein (immunohistochemical) markers of regeneration, neurodegeneration, myogenesis, cell cycle regulation, and inflammation. Our results show morphological changes indicative of active denervation and reinnervation and an increase in small atrophic fibres. We demonstrate differences between ALS and controls in pathways controlling skeletal muscle homeostasis, cytoskeletal and regenerative markers, neurodegenerative factors, myogenic factors, cell cycle determinants, and inflammatory markers. Our results on Pax7 and MyoD protein expression suggest that proliferation and differentiation of skeletal muscle stem cells are affected in ALS patients, and the myogenic processes cannot overcome the denervation-induced wasting.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Slow) antibody produced in mouse, clone NOQ7.5.4D, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid