Merck
E4orf4, a novel adenovirus death factor that induces p53-independent apoptosis by a pathway that is not inhibited by zVAD-fmk.
The Journal of cell biology (1998-03-14)
J N Lavoie, M Nguyen, R C Marcellus, P E Branton, G C Shore
ABSTRAKT
In the absence of E1B, the 289-amino acid product of human adenovirus type 5 13S E1A induces p53-independent apoptosis by a mechanism that requires viral E4 gene products (Marcellus, R.C., J.C. Teodoro, T. Wu, D.E. Brough, G. Ketner, G.C. Shore, and P.E. Branton. 1996. J. Virol. 70:6207-6215) and involves a mechanism that includes activation of caspases (Boulakia, C.A., G. Chen, F.W. Ng, J. G. Teodoro, P.E. Branton, D.W. Nicholson, G.G. Poirier, and G.C. Shore. 1996. Oncogene. 12:529-535). Here, we show that one of the E4 products, E4orf4, is highly toxic upon expression in rodent cells regardless of the p53 status, and that this cytotoxicity is significantly overcome by coexpression with either Bcl-2 or Bcl-XL. Conditional expression of E4orf4 induces a cell death process that is characterized by apoptotic hallmark features, such as externalization of phosphatidylserine, loss of mitochondrial membrane potential, cytoplasmic vacuolation, condensation of chromatin, and internucleosomal DNA degradation. However, the wide-spectrum inhibitor of caspases, tetrapeptide zVAD-fmk, does not affect any of these apoptogenic manifestations, and does not alter the kinetics of E4orf4-induced cell death. Moreover, E4orf4 expression does not result in activation of the downstream effector caspase common to most apoptosis-inducing events, caspase-3 (CPP32). We conclude, therefore, that in the absence of E1A, E4orf4 is sufficient by itself to trigger a p53-independent apoptosis pathway that may operate independently of the known zVAD-inhibitable caspases, and that may involve an as yet uncharacterized mechanism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-Fas (ludzkie, aktywujące), klon CH11, clone CH11, Upstate®, from mouse