Merck
  • Early combined treatment with sildenafil and adipose-derived mesenchymal stem cells preserves heart function in rat dilated cardiomyopathy.
Early combined treatment with sildenafil and adipose-derived mesenchymal stem cells preserves heart function in rat dilated cardiomyopathy.
Journal of translational medicine (2010-09-28)
Yu-Chun Lin, Steve Leu, Cheuk-Kwan Sun, Chia-Hung Yen, Ying-Hsien Kao, Li-Teh Chang, Tzu-Hsien Tsai, Sarah Chua, Morgan Fu, Sheung-Fat Ko, Chiung-Jen Wu, Fan-Yen Lee, Hon-Kan Yip
ABSTRAKT
We investigated whether early combined autologous adipose-derived mesenchymal stem cell (ADMSC) and sildenafil therapy offers an additive benefit in preserving heart function in rat dilated cardiomyopathy (DCM). Adult Lewis rats (n = 8 per group) were divided into group 1 (normal control), group 2 (saline-treated DCM rats), group 3 [2.0 × 106 ADMSC implanted into left ventricular (LV) myocardium of DCM rats], group 4 (DCM rats with sildenafil 30 mg/kg/day, orally), and group 5 (DCM rats with combined ADMSC-sildenafil). Treatment was started 1 week after DCM induction and the rats were sacrificed on day 90. The results showed that mitochondrial protein expressions of connexin43 and cytochrome-C were lowest in group 2, and lower in groups 3 and 4 than in group 5 (p < 0.002). Conversely, oxidative index was highest in group 2, and also higher in groups 3 and 4 than in group 5 (p < 0.0003). The mRNA expressions of interleukin (IL)-10, Gro/IL-8, endothelial nitric oxide synthase, and Bcl-2 were lowest in group 2, and lower in groups 3 and 4 compared with group 5 (p < 0.0001). The mRNA expressions of matrix metalloproteinase-9, Bax, caspase 3, and stromal-cell derived factor-1α were highest in group 2, and higher in groups 3 and 4 than in group 5 (p < 0.0004). Apoptosis and fibrosis in LV myocardium were most prominent in group 2 and higher in groups 3 and 4 than in group 5, whereas angiogenesis and LV ejection fraction were lowest in group 2 and lower in groups 3 and 4 than in group 5 (p < 0.003). Early combined ADMSC/sildenafil is superior to either treatment alone in preserving LV function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.