- MiR-138 downregulates miRNA processing in HeLa cells by targeting RMND5A and decreasing Exportin-5 stability.
MiR-138 downregulates miRNA processing in HeLa cells by targeting RMND5A and decreasing Exportin-5 stability.
Nucleic acids research (2013-09-24)
Jie Li, Ying Chen, Xingliang Qin, Junzhi Wen, Hongmei Ding, Wei Xia, Shaohua Li, Xueting Su, Wei Wang, Hui Li, Qiang Zhao, Tao Fang, Lianghu Qu, Ningsheng Shao
PMID24057215
ABSTRAKT
MicroRNAs (miRNAs) are a class of non-coding small RNAs that consist of ≈ 22 nt and are involved in several biological processes by regulating target gene expression. MiR-138 has many biological functions and is often downregulated in cancers. Our results showed that overexpression of miR-138 downregulated target RMND5A (required for meiotic nuclear division 5 homolog A) and reduced Exportin-5 stability, which results in decreased levels of pre-miRNA nuclear export in HeLa cells. We also found that miR-138 could significantly inhibit HeLa cell migration by targeting RMND5A. Our study therefore identifies miR-138-RMND5A-Exportin-5 as a previously unknown miRNA processing regulatory pathway in HeLa cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-XPO5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Anti-XPO5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution