Merck
Metabolites of cerebellar neurons and hippocampal neurons play opposite roles in pathogenesis of Alzheimer's disease.
PloS one (2009-05-14)
Jing Du, Bing Sun, Kui Chen, Lang Zhang, Shubo Liu, Qingquan Gu, Li Fan, Nanming Zhao, Zhao Wang
ABSTRAKT
Metabolites of neural cells, is known to have a significant effect on the normal physiology and function of neurons in brain. However, whether they play a role in pathogenesis of neurodegenerative diseases is unknown. Here, we show that metabolites of neurons play essential role in the pathogenesis of Alzheimer's disease (AD). Firstly, in vivo and in vitro metabolites of cerebellar neurons both significantly induced the expression of Abeta-degrading enzymes in the hippocampus and cerebral cortex and promoted Abeta clearance. Moreover, metabolites of cerebellar neurons significantly reduced brain Abeta levels and reversed cognitive impairments and other AD-like phenotypes of APP/PS1 transgenic mice, in both early and late stages of AD pathology. On the other hand, metabolites of hippocampal neurons reduced the expression of Abeta-degrading enzymes in the cerebellum and caused cerebellar neurodegeneration in APP/PS1 transgenic mice. Thus, we report, for the first time, that metabolites of neurons not only are required for maintaining the normal physiology of neurons but also play essential role in the pathogenesis of AD and may be responsible for the regional-specificity of Abeta deposition and AD pathology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Ethyl-2-pyrrolidone, 98%