Merck
  • Trimethylamine-N-oxide switches from stabilizing nature: A mechanistic outlook through experimental techniques and molecular dynamics simulation.
Trimethylamine-N-oxide switches from stabilizing nature: A mechanistic outlook through experimental techniques and molecular dynamics simulation.
Scientific reports (2016-03-31)
Anjeeta Rani, Abhilash Jayaraj, B Jayaram, Venkatesu Pannuru
ABSTRAKT
In adaptation biology of the discovery of the intracellular osmolytes, the osmolytes are found to play a central role in cellular homeostasis and stress response. A number of models using these molecules are now poised to address a wide range of problems in biology. Here, a combination of biophysical measurements and molecular dynamics (MD) simulation method is used to examine the effect of trimethylamine-N-oxide (TMAO) on stem bromelain (BM) structure, stability and function. From the analysis of our results, we found that TMAO destabilizes BM hydrophobic pockets and active site as a result of concerted polar and non-polar interactions which is strongly evidenced by MD simulation carried out for 250 ns. This destabilization is enthalpically favourable at higher concentrations of TMAO while entropically unfavourable. However, to the best of our knowledge, the results constitute first detailed unambiguous proof of destabilizing effect of most commonly addressed TMAO on the interactions governing stability of BM and present plausible mechanism of protein unfolding by TMAO.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bromelain from pineapple stem, ≥3 units/mg protein
Sigma-Aldrich
(1S)-(+)-10-Camphorsulfonic acid, 99%