Merck
GPR39 is coupled to TMEM16A in intestinal fibroblast-like cells.
PloS one (2012-11-08)
Fanning Zeng, Nicholas Wind, Conor McClenaghan, J Martin Verkuyl, Robert P Watson, Mark S Nash
ABSTRAKT
GPR39 is a GPCR implicated as a regulator of gastrointestinal motility, although the mechanism remains elusive. Here, we report that GPR39 is expressed by a specific cell population cultured from mouse small intestine muscle layers, which was subsequently identified as fibroblast-like cells (FLCs) that have recently been shown to modulate gut motility. Application of the GPR39 agonist, Zn(2+), induced large currents and membrane depolarization in FLCs cultured from wild-type mice, but not Gpr39(-/-) mice. This Zn(2+)-induced current could be suppressed by application of a TMEM16A antagonist, CaCC(inh)-A01, or by silencing Tmem16a expression. These data suggest that GPR39 might modulate gut motility via regulating TMEM16A function in FLCs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human ANO1