Merck
Clinicopathological significance of expression of p-c-Jun, TCF4 and beta-Catenin in colorectal tumors.
BMC cancer (2008-11-11)
Kayoko Takeda, Ichiro Kinoshita, Yasushi Shimizu, Yusuke Ohba, Tomoo Itoh, Yoshihiro Matsuno, Toshiaki Shichinohe, Hirotoshi Dosaka-Akita
ABSTRAKT
A recent study has shown that phosphorylated c-Jun (p-c-Jun) interacts with TCF4 to form a complex that cooperatively enhances their transcriptional activity in the presence of beta-Catenin, and that their interaction is critical for mouse intestinal tumorigenesis. To determine the significance of these three proteins in human colorectal tumors, we analyzed their nuclear expression by immunohistochemistry. we analyzed their nuclear expression by immunohistochemistry using paraffin-embedded specimens of 68 resected colorectal tumors, which consisted of 19 adenomas, 14 high-grade intraepithelial neoplasia (HGINs) and 35 adenocarcinomas. We also analyzed the expression of MMP7, which has functional AP-1 and TCF binding sites in its promoter. Expression of p-c-Jun, TCF4 and beta-Catenin were significantly higher in adenomas than in the adjacent normal epithelia. Expression of p-c-Jun and beta-Catenin in HGINs and adenocarcinomas were also significantly higher than in the adjacent normal epithelia. p-c-Jun expression, but not TCF4 and beta-Catenin, was higher in adenomas and HGINs than in adenocarcinomas, in which p-c-Jun expression was negatively correlated with pT stage progression. Furthermore, significant correlations of expression were observed between p-c-Jun and TCF4 (r = 0.25, p = 0.04), TCF4 and beta-Catenin (r = 0.30, p = 0.01), p-c-Jun and MMP7 (r = 0.26, p = 0.03), and TCF4 and MMP7 (r = 0.39, p = 0.0008), respectively. These results suggest that nuclear expression of p-c-Jun, TCF4 and beta-Catenin have important roles in human colorectal tumor development and that p-c-Jun may play a pivotal role in the earlier stages of tumor development.