Merck
Expression and function of Allergin-1 on human primary mast cells.
PloS one (2013-10-12)
Kei Nagai, Satoko Tahara-Hanaoka, Yuko Morishima, Takahiro Tokunaga, Yoshimasa Imoto, Emiko Noguchi, Kazumasa Kanemaru, Masamichi Imai, Shiro Shibayama, Nobuyuki Hizawa, Shigeharu Fujieda, Kunihiro Yamagata, Akira Shibuya
ABSTRAKT
Mast cells (MC) play an important role in allergic and non-allergic immune responses. Activation of human MC is modulated by several cell surface inhibitory receptors, including recently identified Allergin-1 expressed on both human and mouse MC. Although Allergin-1 suppresses IgE-mediated, mast cell-dependent anaphylaxis in mice, the expression profile and function of Allergin-1 on human primary MC remains undetermined. Here, we established a seven-color flow cytometry method for assessing expression and function of a very small number of human primary MC. We show that Allergin-1S1, a splicing isoform of Allergin-1, is predominantly expressed on human primary MC in both bronchoalveolar lavage (BAL) fluid and nasal scratching specimens. Moreover, Allergin-1S1 inhibits IgE-mediated activation from human primary MC in BAL fluid. These results indicate that Allergin-1 on human primary MC exhibits similar characteristics as mouse Allergin-1 in the expression profile and function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TNP, ≥95% (HPLC)