Merck
Discovery of SMAD4 promoters, transcription factor binding sites and deletions in juvenile polyposis patients.
Nucleic acids research (2011-03-23)
Daniel Calva, Fadi S Dahdaleh, George Woodfield, Ronald J Weigel, Jennifer C Carr, Sathivel Chinnathambi, James R Howe
ABSTRAKT
Inactivation of SMAD4 has been linked to several cancers and germline mutations cause juvenile polyposis (JP). We set out to identify the promoter(s) of SMAD4, evaluate their activity in cell lines and define possible transcription factor binding sites (TFBS). 5'-rapid amplification of cDNA ends (5'-RACE) and computational analyses were used to identify candidate promoters and corresponding TFBS and the activity of each was assessed by luciferase vectors in different cell lines. TFBS were disrupted by site-directed mutagenesis (SDM) to evaluate the effect on promoter activity. Four promoters were identified, two of which had significant activity in several cell lines, while two others had minimal activity. In silico analysis revealed multiple potentially important TFBS for each promoter. One promoter was deleted in the germline of two JP patients and SDM of several sites led to significant reduction in promoter activity. No mutations were found by sequencing this promoter in 65 JP probands. The predicted TFBS profiles for each of the four promoters shared few transcription factors in common, but were conserved across several species. The elucidation of these promoters and identification of TFBS has important implications for future studies in sporadic tumors from multiple sites, and in JP patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ChIPAb+ RNA Pol II - przeciwciało i zestaw primerów zweryfikowane pod kątem ChIP, from mouse