Merck
Expression of phospho-ERK1/2 and PI3-K in benign and malignant gallbladder lesions and its clinical and pathological correlations.
Journal of experimental & clinical cancer research : CR (2009-05-19)
Qinglong Li, Zhulin Yang
ABSTRAKT
An increasing number of studies have shown that ERK and PI3-K/AKT signaling pathways are involved in various human cancers including hepatocellular carcinoma and cholangiocarcinoma. However, few studies have examined gallbladder cancer specimens, and little is known about the clinical and pathological significance of ERK1/2 and PI3-K/AKT signaling changes in gallbladder adenocarcinoma. In this study, we examined phospho-ERK1/2 (p-ERK1/2) and PI3K expression and analyzed its clinicopathological impact in gallbladder adenocarcinoma. Immunohistochemistry was used to detect and compare the frequency of p-ERK1/2 and PI3-K expression in gallbladder adenocarcinoma, peri-tumor tissues, adenomatous polyps, and chronic cholecystitis specimens. The positive staining for p-EKR1/2 and PI3-K were 63/108 (58.3%) and 55/108 (50.9%) in gallbladder adenocarcinoma; 14/46 (30.4%) and 5/46 (10.1%) in peri-tumor tissues; 3/15 (20%) and 3/15 (20%) in adenomatous polyps; and 4/35 (11.4%) and 3/35 (8.6%) in chronic cholecystitis. The positive rate of p-ERK1/2 or PI3-K in gallbladder adenocarcinoma was significantly higher than that in peri-tumor tissue (both, P < 0.01), adenomatous polyps (p-ERK1/2, P < 0.01; PI3-K, P < 0.05), and chronic cholecystitis (both, P < 0.01). The positive staining for p-ERK1/2 or PI3-K was significantly lower in well/highly-differentiated adenocarcinomas with maximal diameter < 2.0 cm, no metastasis to lymph node, and no infiltration of regional tissues or organs compared to poorly-differentiated adenocarcinomas which are characterized by a maximal diameter >or= 2.0 cm, with metastasis to lymph node and infiltration of regional tissues or organs (P < 0.05 or P < 0.01). Moreover, the frequency of p-ERK1/2 expression in gallbladder adenocarcinomas without gallstone was significantly lower than those with gallstones. Increased expression of p-ERK1/2 (P < 0.05) and PI3K (P = 0.062) was associated with decreased overall survival. Multivariate Cox regression analysis showed that increased p-ERK1/2 expression was an independent prognostic predictor in gallbladder carcinoma (P = 0.028). Increased expression of p-ERK1/2 and PI3K might contribute to gallbladder carcinogenesis. p-ERK1/2 over-expression is correlated with decreased survival and therefore may serve as an important biological marker in development of gallbladder adenocarcinoma.