Merck
Numb regulates cell-cell adhesion and polarity in response to tyrosine kinase signalling.
The EMBO journal (2009-07-18)
Zezhou Wang, Shelley Sandiford, Chenggang Wu, Shawn Shun-Cheng Li
ABSTRAKT
Epithelial-mesenchymal transition (EMT), which can be caused by aberrant tyrosine kinase signalling, marks epithelial tumour progression and metastasis, yet the underlying molecular mechanism is not fully understood. Here, we report that Numb interacts with E-cadherin (E-cad) through its phosphotyrosine-binding domain (PTB) and thereby regulates the localization of E-cad to the lateral domain of epithelial cell-cell junction. Moreover, Numb engages the polarity complex Par3-aPKC-Par6 by binding to Par3 in polarized Madin-Darby canine kidney cells. Intriguingly, after Src activation or hepatocyte growth factor (HGF) treatment, Numb decouples from E-cad and Par3 and associates preferably with aPKC-Par6. Binding of Numb to aPKC is necessary for sequestering the latter in the cytosol during HGF-induced EMT. Knockdown of Numb by small hairpin RNA caused a basolateral-to-apicolateral translocation of E-cad and beta-catenin accompanied by elevated actin polymerization, accumulation of Par3 and aPKC in the nucleus, an enhanced sensitivity to HGF-induced cell scattering, a decrease in cell-cell adhesion, and an increase in cell migration. Our work identifies Numb as an important regulator of epithelial polarity and cell-cell adhesion and a sensor of HGF signalling or Src activity during EMT.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało przeciw fosfotyrozynie, klon ®, clone 4G10®, Upstate®, from mouse