Merck
The Electronic and Magnetic Structure of bcc Iron-Nickel Alloy
Journal of Physics. Condensed Matter : An Institute of Physics Journal (1999)
M. E. Elzain, A.A. Yousif, A.D. Al Rawas, A.M. Gismelseed