Merck
Tackling obstacles for gene therapy targeting neurons: disrupting perineural nets with hyaluronidase improves transduction.
PloS one (2013-01-10)
Klaus Wanisch, Stjepana Kovac, Stephanie Schorge
ABSTRAKT
Gene therapy has been proposed for many diseases in the nervous system. In most cases for successful treatment, therapeutic vectors must be able to transduce mature neurons. However, both in vivo, and in vitro, where preliminary characterisation of viral particles takes place, transduction of neurons is typically inefficient. One possible explanation is that the extracellular matrix (ECM), forming dense perineural nets (PNNs) around neurons, physically blocks access to the cell surface. We asked whether co-administration of lentiviral vectors with an enzyme that disrupts the ECM could improve transduction efficiency. Using hyaluronidase, an enzyme which degrades hyaluronic acid, a high molecular weight molecule of the ECM with mainly a scaffolding function, we show that in vitro in mixed primary cortical cultures, and also in vivo in rat cortex, hyaluronidase co-administration increased the percentage of transduced mature, NeuN-positive neurons. Moreover, hyaluronidase was effective at doses that showed no toxicity in vitro based on propidium iodide staining of treated cultures. Our data suggest that limited efficacy of neuronal transduction is partly due to PNNs surrounding neurons, and further that co-applying hyaluronidase may benefit applications where efficient transduction of neurons in vitro or in vivo is required.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse