Merck
  • Kinesin-1 Expressed in Insect Cells Improves Microtubule in Vitro Gliding Performance, Long-Term Stability and Guiding Efficiency in Nanostructures.
Kinesin-1 Expressed in Insect Cells Improves Microtubule in Vitro Gliding Performance, Long-Term Stability and Guiding Efficiency in Nanostructures.
IEEE transactions on nanobioscience (2016-02-18)
Till Korten, Samata Chaudhuri, Elena Tavkin, Marcus Braun, Stefan Diez
ABSTRAKT
The cytoskeletal motor protein kinesin-1 has been successfully used for many nanotechnological applications. Most commonly, these applications use a gliding assay geometry where substrate-attached motor proteins propel microtubules along the surface. So far, this assay has only been shown to run undisturbed for up to 8 h. Longer run times cause problems like microtubule shrinkage, microtubules getting stuck and slowing down. This is particularly problematic in nanofabricated structures where the total number of microtubules is limited and detachment at the structure walls causes additional microtubule loss. We found that many of the observed problems are caused by the bacterial expression system, which has so far been used for nanotechnological applications of kinesin-1. We strive to enable the use of this motor system for more challenging nanotechnological applications where long-term stability and/or reliable guiding in nanostructures is required. Therefore, we established the expression and purification of kinesin-1 in insect cells which results in improved purity and--more importantly--long-term stability > 24 h and guiding efficiencies of > 90% in lithographically defined nanostructures.