Merck
  • Ferric nitrilotriacetate (Fe-NTA)-induced reactive oxidative species protects human hepatic stellate cells from apoptosis by regulating Bcl-2 family proteins and mitochondrial membrane potential.
Ferric nitrilotriacetate (Fe-NTA)-induced reactive oxidative species protects human hepatic stellate cells from apoptosis by regulating Bcl-2 family proteins and mitochondrial membrane potential.
International journal of clinical and experimental medicine (2016-01-16)
Mei Liu, Shu-Jie Li, Yong-Ning Xin, Shu-Sheng Ji, Rui-Jin Xie, Shi-Ying Xuan
ABSTRAKT
Reactive oxidative species (ROS)-induced apoptosis of human hepatic stellate (HSC) is one of the treatments for liver fibrosis. However, how ROS (reactive oxygen species) affect HSC apoptosis and liver fibrosis is still unknown. In our study, ROS in human HSC cell line LX-2 was induced by ferric nitrilotriacetate (Fe-NTA) and assessed by superoxide dismutase (SOD) activity and methane dicarboxylic aldehyde (MDA) level. We found that in LX2 cells Fe-NTA induced notable ROS, which played a protective role in HSCs cells apoptosis by inhibiting Caspase-3 activation. Fe-NTA-induced ROS increased mRNA and protein level of anti-apoptosis Bcl-2 and decreased mRNA protein level of pro-apoptosis gene Bax, As a result, maintaining mitochondrial membrane potential of HSCs. Fe-NTA-induced ROS play a protective role in human HSCs by regulating Bcl-2 family proteins and mitochondrial membrane potential.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)