Merck
  • Selective extraction of intracellular components from the microalga Chlorella vulgaris by combined pulsed electric field-temperature treatment.
Selective extraction of intracellular components from the microalga Chlorella vulgaris by combined pulsed electric field-temperature treatment.
Bioresource technology (2016-01-02)
P R Postma, G Pataro, M Capitoli, M J Barbosa, R H Wijffels, M H M Eppink, G Olivieri, G Ferrari
ABSTRAKT
The synergistic effect of temperature (25-65 °C) and total specific energy input (0.55-1.11 kWh kgDW(-1)) by pulsed electric field (PEF) on the release of intracellular components from the microalgae Chlorella vulgaris was studied. The combination of PEF with temperatures from 25 to 55 °C resulted in a conductivity increase of 75% as a result of cell membrane permeabilization. In this range of temperatures, 25-39% carbohydrates and 3-5% proteins release occurred and only for carbohydrate release a synergistic effect was observed at 55 °C. Above 55 °C spontaneous cell lysis occurred without PEF. Combined PEF-temperature treatment does not sufficiently disintegrate the algal cells to release both carbohydrates and proteins at yields comparable to the benchmark bead milling (40-45% protein, 48-58% carbohydrates).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%