Merck
  • Over-expression of CYP6A2 is associated with spirotetramat resistance and cross-resistance in the resistant strain of Aphis gossypii Glover.
Over-expression of CYP6A2 is associated with spirotetramat resistance and cross-resistance in the resistant strain of Aphis gossypii Glover.
Pesticide biochemistry and physiology (2016-01-19)
Tianfei Peng, Yiou Pan, Chen Yang, Xiwu Gao, Jinghui Xi, Yongqiang Wu, Xiao Huang, E Zhu, Xuecheng Xin, Chao Zhan, Qingli Shang
ABSTRAKT
A laboratory-selected spirotetramat-resistant strain (SR) of cotton aphid developed 579-fold and 15-fold resistance to spirotetramat in adult aphids and 3rd instar nymphs, respectively, compared with a susceptible strain (SS) [26]. The SR strain developed high-level cross-resistance to alpha-cypermethrin and bifenthrin and very low or no cross-resistance to the other tested insecticides. Synergist piperonyl butoxide (PBO) dramatically increased the toxicity of spirotetramat and alpha-cypermethrin in the resistant strain. RT-qPCR results demonstrated that the transcriptional levels of CYP6A2 increased significantly in the SR strain compared with the SS strain, which was consistent with the transcriptome results [30]. The depletion of CYP6A2 transcripts by RNAi also significantly increased the sensitivity of the resistant aphid to spirotetramat and alpha-cypermethrin. These results indicate the possible involvement of CYP6A2 in spirotetramat resistance and alpha-cypermethrin cross-resistance in the cotton aphid. These together with other cross-resistance results have implications for the successful implementation of resistance management strategies for Aphis gossypii.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Spirotetramat, PESTANAL®, analytical standard