Merck
HRMS Profile of a Hazelnut Skin Proanthocyanidin-rich Fraction with Antioxidant and Anti-Candida albicans Activities.
Journal of agricultural and food chemistry (2016-01-08)
Anna Lisa Piccinelli, Imma Pagano, Tiziana Esposito, Teresa Mencherini, Amalia Porta, Anna Maria Petrone, Patrizia Gazzerro, Patrizia Picerno, Francesca Sansone, Luca Rastrelli, Rita Patrizia Aquino
ABSTRAKT
Roasted hazelnut skins (RHS) represent a byproduct of kernel industrial processing. In this research, a RHS extract (RHS-M) and its fraction RHS-M-F3 enriched in proanthocyanidins (PAs), with antioxidant activity, were characterized in terms of total phenolic compound and PA contents. RHS-M and RHS-M-F3 showed antifungal properties against Candida albicans SC5314 (MIC2 = 3.00 and 0.10 μg/mL and MIC0 = 5.00 and 0.50 μg/mL, respectively), determined by the microbroth dilution method and Candida albicans morphological analysis. No cytotoxic effect on HEKa and HDFa cell lines was exhibited by RHS-M and RHS-M-F3. The metabolite profiling of RHS-M and RHS-M-F3 was performed by thiolysis followed by HPLC-UV-HRMS analysis and a combination of HRMS-FIA and HPLC-HRMS(n). Extract and fraction contain oligomeric PAs (mDP of 7.3 and 6.0, respectively, and DP up to 10) mainly constituted by B-type oligomers of (epi)-catechin. Also, (epi)-gallocatechin and gallate derivatives were identified as monomer units, and A-type PAs were detected as minor compounds.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Procyanidin B2, analytical standard
Millipore
DMACA Reagent, suitable for microbiology