Merck
  • Presence of ?-glucanase activity in wheat and dairy ingredients and use of organic salts as potential enzyme inhibitors.
    Tosh SM, et al.
Presence of ?-glucanase activity in wheat and dairy ingredients and use of organic salts as potential enzyme inhibitors.<BR/> Tosh SM, et al.
Journal of Cereal Science (2012)
Tosh SM, et al.