Merck
Effects of intrinsic aerobic capacity, aging and voluntary running on skeletal muscle sirtuins and heat shock proteins.
Experimental gerontology (2016-04-04)
Sira Karvinen, Mika Silvennoinen, Petra Vainio, Lea Sistonen, Lauren G Koch, Steven L Britton, Heikki Kainulainen
ABSTRAKT
Sirtuins are proteins that connect energy metabolism, oxidative stress and aging. Expression of heat shock proteins (Hsps) is regulated by heat shock factors (HSFs) in response to various environmental and physiological stresses, such as oxidative stress. Oxidative stress accumulates during aging which makes cells more prone to DNA damage. Although many experimental animal models have been designed to study the effects of knockdown or overexpression of sirtuins, HSFs and Hsps, little is known about how aging per se affects their expression. Here we study the impact of intrinsic aerobic capacity, aging and voluntary exercise on the levels of sirtuins, HSFs and Hsps in skeletal muscle. We studied the protein levels of sirtuins (SIRT1, SIRT2, SIRT3, SIRT4, SIRT5, SIRT6 and SIRT7), HSF1, HSF2, Hsp10, Hsp27 and Hsp70 before and after one-year of voluntary running intervention of rat strains selectively bred for intrinsic aerobic exercise capacity; high capacity runners (HCR) and low capacity runners (LCR) differ by more than 30% for median lifespan. This setup enabled us to discern the effects of inborn aerobic capacity, aging and exercise activity on the protein levels of sirtuins, HSFs and Hsps in skeletal muscle. Our results revealed that the longer lived HCR rats had higher SIRT3, HSF1 and HSF2 contents in skeletal muscle (gastrocnemius, p < 0.05) than LCRs. Neither aging nor voluntary running had a significant effect on the studied sirtuin proteins. Aging significantly increased the protein levels of HSF1, HSF2 and Hsp27 (p < 0.05). Our finding of elevated SIRT3 levels in HCR rats is in line with previous studies; SIRT3 in general is linked to elevated fatty acid oxidation and oxidative phosphorylation, which previously have been associated with metabolic profile of HCRs. HSF1, HSF2 and Hsp27 levels increased with aging, showing that aged muscles responded to aging-related stress. Our study shows for the first time that SIRT3 protein level is linked to high inborn aerobic capacity, and may be directly interconnected to longevity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-HSP10 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture