- A new type of soluble pentacene precursor for organic thin-film transistors.
A new type of soluble pentacene precursor for organic thin-film transistors.
Chemical communications (Cambridge, England) (2007-02-28)
Kew-Yu Chen, Hsing-Hung Hsieh, Chung-Chih Wu, Jiunn-Jye Hwang, Tahsin J Chow
PMID17325807
ABSTRAKT
A new type of soluble pentacene precursor is synthesized, which extrudes a unit of CO upon heating at 150 degrees C, to produce pentacene in nearly quantitative yield.