- Rhenium-catalyzed insertion of aldehyde into a C-H bond: synthesis of isobenzofuran derivatives.
Rhenium-catalyzed insertion of aldehyde into a C-H bond: synthesis of isobenzofuran derivatives.
Journal of the American Chemical Society (2006-09-21)
Yoichiro Kuninobu, Yuta Nishina, Chie Nakagawa, Kazuhiko Takai
PMID16984162
ABSTRAKT
A rhenium complex, [ReBr(CO)3(thf)]2, catalyzed reactions of aromatic ketimines with aldehydes to give isobenzofuran derivatives in good to excellent yields. In contrast to ruthenium and rhodium catalysts, aldehydes, which are polar unsaturated molecules, inserted into the C-H bond after activation by the rhenium complex.