Merck
Trifluoroselenomethionine: A New Unnatural Amino Acid.
Chembiochem : a European journal of chemical biology (2016-07-08)
Eric Block, Squire J Booker, Sonia Flores-Penalba, Graham N George, Sivaji Gundala, Bradley J Landgraf, Jun Liu, Stephene N Lodge, M Jake Pushie, Sharon Rozovsky, Abith Vattekkatte, Rama Yaghi, Huawei Zeng
ABSTRAKT
Trifluoroselenomethionine (TFSeM), a new unnatural amino acid, was synthesized in seven steps from N-(tert-butoxycarbonyl)-l-aspartic acid tert-butyl ester. TFSeM shows enhanced methioninase-induced cytotoxicity, relative to selenomethionine (SeM), toward HCT-116 cells derived from human colon cancer. Mechanistic explanations for this enhanced activity are computationally and experimentally examined. Comparison of TFSeM and SeM by selenium EXAFS and DFT calculations showed them to be spectroscopically and structurally very similar. Nonetheless, when two different variants of the protein GB1 were expressed in an Escherichia coli methionine auxotroph cell line in the presence of TFSeM and methionine (Met) in a 9:1 molar ratio, it was found that, surprisingly, 85 % of the proteins contained SeM residues, even though no SeM had been added, thus implying loss of the trifluoromethyl group from TFSeM. The transformation of TFSeM into SeM is enzymatically catalyzed by E. coli extracts, but TFSeM is not a substrate of E. coli methionine adenosyltransferase.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Benzamidine, ≥95.0%