Merck
  • Reactivity, Selectivity, and Reaction Mechanisms of Aminoguanidine, Hydralazine, Pyridoxamine, and Carnosine as Sequestering Agents of Reactive Carbonyl Species: A Comparative Study.
Reactivity, Selectivity, and Reaction Mechanisms of Aminoguanidine, Hydralazine, Pyridoxamine, and Carnosine as Sequestering Agents of Reactive Carbonyl Species: A Comparative Study.
ChemMedChem (2016-02-19)
Mara Colzani, Danilo De Maddis, Gaia Casali, Marina Carini, Giulio Vistoli, Giancarlo Aldini
ABSTRAKT
Reactive carbonyl species (RCS) are endogenous or exogenous byproducts involved in the pathogenic mechanisms of different oxidative-based disorders. Detoxification of RCS by carbonyl quenchers is a promising therapeutic strategy. Among the most studied quenchers are aminoguanidine, hydralazine, pyridoxamine, and carnosine; their quenching activity towards four RCS (4-hydroxy-trans-2-nonenal, methylglyoxal, glyoxal, and malondialdehyde) was herein analyzed and compared. Their ability to prevent protein carbonylation was evaluated in vitro by using an innovative method based on high-resolution mass spectrometry (HRMS). The reactivity of the compounds was RCS dependent: carnosine efficiently quenched 4-hydroxy-trans-2-nonenal, pyridoxamine was particularly active towards malondialdehyde, aminoguanidine was active towards methylglyoxal and glyoxal, and hydralazine efficiently quenched all RCS. Reaction products were generated in vitro and were characterized by HRMS. Molecular modeling studies revealed that the reactivity was controlled by specific stereoelectronic parameters that could be used for the rational design of improved carbonyl quenchers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ubiquitin from bovine erythrocytes, BioUltra, ≥98% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Hydralazine hydrochloride