Merck
Antagonistic roles of Drosophila Tctp and Brahma in chromatin remodelling and stabilizing repeated sequences.
Nature communications (2016-10-01)
Sung-Tae Hong, Kwang-Wook Choi
ABSTRAKT
Genome stability is essential for all organisms. Translationally controlled tumour protein (TCTP) is a conserved protein associated with cancers. TCTP is involved in multiple intracellular functions, but its role in transcription and genome stability is poorly understood. Here, we demonstrate new functions of Drosophila TCTP (Tctp) in transcription and the stability of repeated sequences (rDNA and pericentromeric heterochromatin). Tctp binds Brahma (Brm) chromatin remodeler to negatively modulate its activity. Tctp mutants show abnormally high levels of transcription in a large set of genes and transposons. These defects are ameliorated by brm mutations. Furthermore, Tctp promotes the stability of repeated sequences by opposing the Brm function. Additional regulation of pericentromeric heterochromatin by Tctp is mediated by su(var)3-9 transcriptional regulation. Altogether, Tctp regulates transcription and the stability of repeated sequences by antagonizing excess Brm activity. This study provides insights into broader nuclear TCTP functions for the maintenance of genome stability.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Przeciwciało anty-histonowe H3, CT, pan, klon A3S, królicze monoklonalne, clone A3S, Upstate®, from rabbit
Sigma-Aldrich
Przeciwciało przeciw dimetylo-histonowi H3 (Lys9), Upstate®, from rabbit
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-V5 antibody produced in rabbit, IgG fraction of antiserum