- Structural elucidation of polysaccharide containing 3-O-methyl galactose from fruiting bodies of Pleurotus citrinopileatus.
Structural elucidation of polysaccharide containing 3-O-methyl galactose from fruiting bodies of Pleurotus citrinopileatus.
Carbohydrate research (2016-10-23)
Pengfei He, Anqiang Zhang, Saijing Zhou, Fuming Zhang, Robert J Linhardt, Peilong Sun
PMID27599128
ABSTRAKT
A water-soluble polysaccharide containing 3-O-methyl galactose (PCP60W) was isolated from fruiting bodies of Pleurotus citrinopileatus and purified by anion-exchange and gel column chromatography. This polysaccharide has an average molecular weight of 2.74 × 10
MATERIAŁY