Merck
Polydiacetylenyl β-cyclodextrin based smart vesicles for colorimetric assay of arginine and lysine.
Scientific reports (2016-08-10)
Eunae Cho, Hwanhee Kim, Youngjin Choi, Seung R Paik, Seunho Jung
ABSTRAKT
Selective visualization of arginine and lysine has been explored among 20 amino acids using the hybrid conjugate of β-cyclodextrin (β-CD) and polydiacetylene (PDA). The mono pentacosa-10,12-diynyl aminomethyl group was successfully coupled to either the primary or the secondary face of β-CD, where mono-6-amino-6-deoxy-β-CD or mono-3-amino-3-deoxy-β-CD reacted with the N-hydroxysuccinimide ester of 10,12-pentacosadiynoic acid. In this combinatorial system, the cylindrical β-cyclodextrin functions as a channel for the introduction of the cationic amino acids to the artificial membrane. The membrane perturbation and aggregation by the target amino acids could be exclusively visualized as a blue to red color change based on the responsive polydiacetylene domain. These interesting findings demonstrated that the developed β-CD conjugated PDA system may offer a new method of cell-penetrating mechanism, a promising vector system, as well as impact the production industry of arginine or lysine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Adamantanecarboxylic acid, 99%