- Effect of surfactants on surface activity and rheological properties of type I collagen at air/water interface.
Effect of surfactants on surface activity and rheological properties of type I collagen at air/water interface.
Colloids and surfaces. B, Biointerfaces (2016-11-05)
Aleksandra Kezwoń, Ilona Góral, Tomasz Frączyk, Kamil Wojciechowski
PMID27616064
ABSTRAKT
We describe the effect of three synthetic surfactants (anionic - sodium dodecyl sulfate (SDS), cationic - cetyltrimethylammonium bromide (CTAB) and nonionic - Triton X-100 (TX-100)) on surface properties of the type I calf skin collagen at the air/water interface in acidic solutions (pH 1.8). The protein concentration was fixed at 5×10
MATERIAŁY