- Functional competence of a partially engaged GPCR-β-arrestin complex.
Functional competence of a partially engaged GPCR-β-arrestin complex.
Nature communications (2016-11-09)
Punita Kumari, Ashish Srivastava, Ramanuj Banerjee, Eshan Ghosh, Pragya Gupta, Ravi Ranjan, Xin Chen, Bhagyashri Gupta, Charu Gupta, Deepika Jaiman, Arun K Shukla
PMID27827372
ABSTRAKT
G Protein-coupled receptors (GPCRs) constitute the largest family of cell surface receptors and drug targets. GPCR signalling and desensitization is critically regulated by β-arrestins (βarr). GPCR-βarr interaction is biphasic where the phosphorylated carboxyl terminus of GPCRs docks to the N-domain of βarr first and then seven transmembrane core of the receptor engages with βarr. It is currently unknown whether fully engaged GPCR-βarr complex is essential for functional outcomes or partially engaged complex can also be functionally competent. Here we assemble partially and fully engaged complexes of a chimeric β
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution