Merck
  • Development of the Larval Amphibian Growth and Development Assay: effects of chronic 4-tert-octylphenol or 17β-trenbolone exposure in Xenopus laevis from embryo to juvenile.
Development of the Larval Amphibian Growth and Development Assay: effects of chronic 4-tert-octylphenol or 17β-trenbolone exposure in Xenopus laevis from embryo to juvenile.
Journal of applied toxicology : JAT (2016-05-05)
Jonathan T Haselman, Patricia A Kosian, Joseph J Korte, Allen W Olmstead, Taisen Iguchi, Rodney D Johnson, Sigmund J Degitz
ABSTRAKT
The Larval Amphibian Growth and Development Assay (LAGDA) is a globally harmonized test guideline developed by the U.S. Environmental Protection Agency in collaboration with Japan's Ministry of the Environment. The LAGDA was designed to evaluate apical effects of chronic chemical exposure on growth, thyroid-mediated amphibian metamorphosis and reproductive development. During the validation phase, two well-characterized endocrine-disrupting chemicals were tested to evaluate the performance of the initial assay design: xenoestrogen 4-tert-octylphenol (tOP) and xenoandrogen 17β-trenbolone (TB). Xenopus laevis embryos were exposed, in flow-through conditions, to tOP (nominal concentrations: 0.0, 6.25, 12.5, 25 and 50 µg l