Merck
Nonstoichiometric acid-base reaction as reliable synthetic route to highly stable CH
Nature communications (2016-11-16)
Mingzhu Long, Tiankai Zhang, Yang Chai, Chun-Fai Ng, Thomas C W Mak, Jianbin Xu, Keyou Yan
ABSTRAKT
Perovskite solar cells have received worldwide interests due to swiftly improved efficiency but the poor stability of the perovskite component hampers the device fabrication under normal condition. Herein, we develop a reliable nonstoichiometric acid-base reaction route to stable perovskite films by intermediate chemistry and technology. Perovskite thin-film prepared by nonstoichiometric acid-base reaction route is stable for two months with negligible PbI
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-tert-Butylpyridine, 98%