Merck
Mechanism and Kinetics of Heterogeneous Reactions of Unsaturated Organic Acids on α-Al
Chemphyschem : a European journal of chemical physics and physical chemistry (2016-08-10)
Lingyan Wu, Qifan Liu, Shengrui Tong, Bo Jing, Weigang Wang, Yucong Guo, Maofa Ge
ABSTRAKT
Heterogeneous reactions have a vital role in the atmosphere due to their significant effects on the evolution of atmospheric aerosols, which in turn contribute to air pollution. However, the mechanism and kinetics of these processes involving unsaturated organic acids, important types of volatile organic compounds, are still unclear. In this work, the heterogeneous uptake of two representative atmospheric unsaturated organic acids (acrylic acid and methacrylic acid) on mineral aerosols including α-Al
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium acrylate, 97%
Sigma-Aldrich
Sodium methacrylate, 99%