Merck
  • Vascular Calcification Induced by Chronic Kidney Disease Is Mediated by an Increase of 1α-Hydroxylase Expression in Vascular Smooth Muscle Cells.
Vascular Calcification Induced by Chronic Kidney Disease Is Mediated by an Increase of 1α-Hydroxylase Expression in Vascular Smooth Muscle Cells.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2016-10-21)
Noelia Torremadé, Milica Bozic, Sara Panizo, Sara Barrio-Vazquez, Jose L Fernandez-Martín, Mario Encinas, David Goltzman, Maria V Arcidiacono, Elvira Fernandez, José M Valdivielso
ABSTRAKT
Vascular calcification (VC) is a complication of chronic kidney disease that predicts morbidity and mortality. Uremic serum promotes VC, but the mechanism involved is unknown. A role for 1,25(OH)
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)