- Recent pause in the growth rate of atmospheric CO
Recent pause in the growth rate of atmospheric CO
Nature communications (2016-11-09)
Trevor F Keenan, I Colin Prentice, Josep G Canadell, Christopher A Williams, Han Wang, Michael Raupach, G James Collatz
PMID27824333
ABSTRAKT
Terrestrial ecosystems play a significant role in the global carbon cycle and offset a large fraction of anthropogenic CO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)