Merck
Vesl/Homer proteins regulate ryanodine receptor type 2 function and intracellular calcium signaling.
Cell calcium (2003-07-31)
Jens H Westhoff, Sung-Yong Hwang, R Scott Duncan, Fumiko Ozawa, Pompeo Volpe, Kaoru Inokuchi, Peter Koulen
ABSTRAKT
Cellular signaling proteins such as metabotropic glutamate receptors, Shank, and different types of ion channels are physically linked by Vesl (VASP/Ena-related gene up-regulated during seizure and LTP)/Homer proteins [Curr. Opin. Neurobiol. 10 (2000) 370; Trends Neurosci. 23 (2000) 80; J. Cell Sci. 113 (2000) 1851]. Vesl/Homer proteins have also been implicated in differentiation and physiological adaptation processes [Nat. Neurosci. 4 (2001) 499; Nature 411 (2001) 962; Biochem. Biophys. Res. Commun. 279 (2000) 348]. Here we provide evidence that a Vesl/Homer subtype, Vesl-1L/Homer-1c (V-1L), reduces the function of the intracellular calcium channel ryanodine receptor type 2 (RyR2). In contrast, Vesl-1S/Homer-1a (V-1S) had no effect on RyR2 function but reversed the effects of V-1L. In live cells, in calcium release studies and in single-channel electrophysiological recordings of RyR2, V-1L reduced RyR2 activity. Important physiological functions and pharmacological properties of RyR2 are preserved in the presence of V-1L. Our findings demonstrate that a protein-protein interaction between V-1L and RyR2 is not only necessary for organizing the structure of intracellular calcium signaling proteins [Curr. Opin. Neurobiol. 10 (2000) 370; Trends Neurosci. 23(2000)80; J. Cell Sci. 113 (2000) 1851; Nat Neurosci. 4 (2001) 499; Nature 411 (2001) 962; Biochem. Biophys. Res. Commun. 279 (2000) 348; Nature 386 (1997) 284], but that V-1L also directly regulates RyR2 channel activity by changing its biophysical properties. Thereby it may control cellular calcium homeostasis. These observations suggest a novel mechanism for the regulation of RyR2 and calcium-dependent cellular functions.