Merck
ThermoMouse: an in vivo model to identify modulators of UCP1 expression in brown adipose tissue.
Cell reports (2014-12-04)
Andrea Galmozzi, Si B Sonne, Svetlana Altshuler-Keylin, Yutaka Hasegawa, Kosaku Shinoda, Ineke H N Luijten, Jae Won Chang, Louis Z Sharp, Benjamin F Cravatt, Enrique Saez, Shingo Kajimura
ABSTRAKT
Obesity develops when energy intake chronically exceeds energy expenditure. Because brown adipose tissue (BAT) dissipates energy in the form of heat, increasing energy expenditure by augmenting BAT-mediated thermogenesis may represent an approach to counter obesity and its complications. The ability of BAT to dissipate energy is dependent on expression of mitochondrial uncoupling protein 1 (UCP1). To facilitate the identification of pharmacological modulators of BAT UCP1 levels, which may have potential as antiobesity medications, we developed a transgenic model in which luciferase activity faithfully mimics endogenous UCP1 expression and its response to physiologic stimuli. Phenotypic screening of a library using cells derived from this model yielded a small molecule that increases UCP1 expression in brown fat cells and mice. Upon adrenergic stimulation, compound-treated mice showed increased energy expenditure. These tools offer an opportunity to identify pharmacologic modulators of UCP1 expression and uncover regulatory pathways that impact BAT-mediated thermogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture