Merck
Hypothalamic leptin action is mediated by histone deacetylase 5.
Nature communications (2016-03-01)
Dhiraj G Kabra, Katrin Pfuhlmann, Cristina García-Cáceres, Sonja C Schriever, Veronica Casquero García, Adam Fiseha Kebede, Esther Fuente-Martin, Chitrang Trivedi, Kristy Heppner, N Henriette Uhlenhaut, Beata Legutko, Uma D Kabra, Yuanqing Gao, Chun-Xia Yi, Carmelo Quarta, Christoffer Clemmensen, Brian Finan, Timo D Müller, Carola W Meyer, Marcelo Paez-Pereda, Kerstin Stemmer, Stephen C Woods, Diego Perez-Tilve, Robert Schneider, Eric N Olson, Matthias H Tschöp, Paul T Pfluger
ABSTRAKT
Hypothalamic leptin signalling has a key role in food intake and energy-balance control and is often impaired in obese individuals. Here we identify histone deacetylase 5 (HDAC5) as a regulator of leptin signalling and organismal energy balance. Global HDAC5 KO mice have increased food intake and greater diet-induced obesity when fed high-fat diet. Pharmacological and genetic inhibition of HDAC5 activity in the mediobasal hypothalamus increases food intake and modulates pathways implicated in leptin signalling. We show HDAC5 directly regulates STAT3 localization and transcriptional activity via reciprocal STAT3 deacetylation at Lys685 and phosphorylation at Tyr705. In vivo, leptin sensitivity is substantially impaired in HDAC5 loss-of-function mice. Hypothalamic HDAC5 overexpression improves leptin action and partially protects against HFD-induced leptin resistance and obesity. Overall, our data suggest that hypothalamic HDAC5 activity is a regulator of leptin signalling that adapts food intake and body weight to our dietary environment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-GFAP antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Informacje o cenach i dostępności nie są obecnie dostępne.